Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARRDC4
Entrez Gene ID
100855829
Gene Name
arrestin domain containing 4
Gene Aliases
-
Chromosome Location
Chr.3: 42650257 - 42663526 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003638967.4 XP_003639015.1 3 - 4 517 119
XM_022416894.1 XP_022272602.1 3 - 4 509 119

View Full Product Details