Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
NRN1L
Entrez Gene ID
611107
Gene Name
neuritin 1 like
Gene Aliases
-
Chromosome Location
Chr.5: 81600442 - 81602175 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005620807.3 XP_005620864.1 2 - 2 405 167
XM_014114074.2 XP_013969549.1 2 - 2 402 167

View Full Product Details