Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHF23
Entrez Gene ID
489465
Gene Name
PHD finger protein 23
Gene Aliases
-
Chromosome Location
Chr.5: 32202636 - 32207738 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005619964.3 XP_005620021.1 4 - 5 1598 98

View Full Product Details