Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PKDCC
Entrez Gene ID
611604
Gene Name
protein kinase domain containing, cytoplasmic
Gene Aliases
-
Chromosome Location
Chr.17: 33659352 - 33669237 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_849285.5 XP_854378.1 1 - 2 678 137

View Full Product Details