Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TBX2
Entrez Gene ID
448779
Gene Name
T-box transcription factor 2
Gene Aliases
-
Chromosome Location
Chr.9: 35237323 - 35245066 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022423257.1 XP_022278965.1 3 - 4 853 69

View Full Product Details