Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TAS2R42
Entrez Gene ID
100271731
Gene Name
taste receptor, type 2, member 42
Gene Aliases
CAFA-T2R55
Chromosome Location
Chr.27: 34739537 - 34740508 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145496.1 NP_001138968.1 1 - 1 851 70

View Full Product Details