Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TLR10
Entrez Gene ID
100379585
Gene Name
toll like receptor 10
Gene Aliases
-
Chromosome Location
Chr.3: 73561596 - 73589286 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001173127.1 NP_001166598.1 1 - 1 2355 100
XM_005618242.3 XP_005618299.1 3 - 3 2687 100

View Full Product Details