Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KRT71
Entrez Gene ID
486522
Gene Name
keratin 71
Gene Aliases
-
Chromosome Location
Chr.27: 2536831 - 2545103 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197029.1 NP_001183958.1 3 - 4 717 73
GenBank mRNA FJ906970.1 - 3 - 4 717 73

View Full Product Details