Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OSTC
Entrez Gene ID
477512
Gene Name
oligosaccharyltransferase complex non-catalytic subunit
Gene Aliases
-
Chromosome Location
Chr.32: 29017421 - 29028119 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160121.1 NP_001153593.1 4 - 4 510 87
GenBank mRNA DN393406.1 - 4 - 4 537 87
DN863568.1 - 1 - 1 519 87

View Full Product Details