Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MIR34B
Entrez Gene ID
100885981
Gene Name
microRNA mir-34b
Gene Aliases
cfa-mir-34b
Chromosome Location
Chr.5: 21558685 - 21558768 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_049425.1 - 48 71

View Full Product Details