Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Thap7
Entrez Gene ID
100753121
Gene Name
THAP domain containing 7
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613919: 1457257 - 1460595 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003502827.2 XP_003502875.1 3 - 4 316 70
XM_007643466.1 XP_007641656.1 3 - 4 255 70
XM_007643467.1 XP_007641657.1 2 - 3 339 70
XM_007643468.1 XP_007641658.1 2 - 3 321 70
GenBank mRNA JI887895.1 - 3 - 4 273 70
JP058954.1 - 3 - 4 273 70

View Full Product Details