Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rbm3
Entrez Gene ID
100689372
Gene Name
RNA binding motif (RNP1, RRM) protein 3
Gene Aliases
I79_012278
Chromosome Location
Chr.Un|NW_003614122: 838784 - 842085 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001246799.1 NP_001233728.1 2 - 3 212 78
XM_007645600.1 XP_007643790.1 3 - 4 330 78
XM_007645602.1 XP_007643792.1 3 - 4 330 78
GenBank mRNA EU556497.1 - 2 - 3 212 78
JI874849.1 - 4 - 5 738 78

View Full Product Details