Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hmga1
Entrez Gene ID
100689040
Gene Name
high mobility group AT-hook 1
Gene Aliases
HMG-I(Y), HMG-Y, HMGIY, Hmg-i
Chromosome Location
Chr.Un|NW_003614227: 709417 - 715192 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244028.1 NP_001230957.1 1 - 2 139 99
XM_007646787.1 XP_007644977.1 2 - 3 344 99
XM_007646788.1 XP_007644978.1 2 - 3 325 99
XM_007646789.1 XP_007644979.1 2 - 3 452 99
XM_007646790.1 XP_007644980.1 2 - 3 486 99
XM_007646791.1 XP_007644981.1 2 - 3 448 99
XM_007646792.1 XP_007644982.1 2 - 3 200 99
XM_007646793.1 XP_007644983.1 2 - 3 199 99
GenBank mRNA AF193762.2 - 1 - 2 139 99

View Full Product Details