Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rps14
Entrez Gene ID
100689292
Gene Name
ribosomal protein S14
Gene Aliases
EMTB
Chromosome Location
Chr.Un|NW_003614229: 654889 - 659378 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244519.1 NP_001231448.1 4 - 5 413 95
XM_007646826.1 XP_007645016.1 3 - 4 488 95
GenBank mRNA JI869879.1 - 4 - 5 462 95
JP040329.1 - 4 - 5 462 95
M11241.1 - 4 - 5 413 95

View Full Product Details