Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pmepa1
Entrez Gene ID
100762220
Gene Name
prostate transmembrane protein, androgen induced 1
Gene Aliases
I79_007840
Chromosome Location
Chr.Un|NW_003613850: 286870 - 294715 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007642095.1 XP_007640285.1 2 - 3 194 103

View Full Product Details