Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Oprd1
Entrez Gene ID
100762884
Gene Name
opioid receptor, delta 1
Gene Aliases
I79_004173
Chromosome Location
Chr.Un|NW_003613689: 2013237 - 2045775 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003498350.1 XP_003498398.1 1 - 2 126 56

View Full Product Details