Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fndc9
Entrez Gene ID
100768078
Gene Name
fibronectin type III domain containing 9
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614829: 405015 - 405695 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510091.1 XP_003510139.1 1 - 1 622 59

View Full Product Details