Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zdbf2
Entrez Gene ID
100756829
Gene Name
zinc finger, DBF-type containing 2
Gene Aliases
I79_008597
Chromosome Location
Chr.Un|NW_003613877: 964468 - 992669 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007642748.1 XP_007640938.1 1 - 2 56 69

View Full Product Details