Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Btbd18
Entrez Gene ID
100754420
Gene Name
BTB (POZ) domain containing 18
Gene Aliases
I79_013442
Chromosome Location
Chr.Un|NW_003614216: 375555 - 384783 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506148.2 XP_003506196.1 3 - 4 432 73

View Full Product Details