Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Shisa3
Entrez Gene ID
100756690
Gene Name
shisa family member 3
Gene Aliases
I79_022375
Chromosome Location
Chr.Un|NW_003615913: 55238 - 62636 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003513408.1 XP_003513456.1 2 - 3 273 70

View Full Product Details