Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nme5
Entrez Gene ID
100774762
Gene Name
NME/NM23 family member 5
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613902: 827103 - 837232 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003502524.2 XP_003502572.1 3 - 4 434 66

View Full Product Details