Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lcn10
Entrez Gene ID
100762990
Gene Name
lipocalin 10
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003617557: 31446 - 34385 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003515217.1 XP_003515265.1 3 - 4 311 80

View Full Product Details