Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nanog
Entrez Gene ID
100774227
Gene Name
Nanog homeobox
Gene Aliases
I79_018432
Chromosome Location
Chr.Un|NW_003614841: 49673 - 53908 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007650806.1 XP_007648996.1 1 - 2 159 80

View Full Product Details