Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Npdc1
Entrez Gene ID
100771936
Gene Name
neural proliferation, differentiation and control, 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003616838: 50298 - 56276 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003514668.2 XP_003514716.1 7 - 8 932 58

View Full Product Details