Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
A3galt2
Entrez Gene ID
100774283
Gene Name
alpha 1,3-galactosyltransferase 2
Gene Aliases
I79_006886
Chromosome Location
Chr.Un|NW_003613801: 961087 - 970729 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003500624.1 XP_003500672.1 2 - 3 102 88

View Full Product Details