Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Snrnp48
Entrez Gene ID
100766215
Gene Name
small nuclear ribonucleoprotein 48kDa (U11/U12)
Gene Aliases
I79_008474
Chromosome Location
Chr.Un|NW_003613871: 796584 - 816402 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003502004.2 XP_003502052.1 4 - 5 432 73

View Full Product Details