Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Il22ra2
Entrez Gene ID
100751510
Gene Name
interleukin 22 receptor, alpha 2
Gene Aliases
I79_001980
Chromosome Location
Chr.Un|NW_003613627: 2201796 - 2213254 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496558.1 XP_003496606.1 3 - 4 375 91

View Full Product Details