Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pcdhga9
Entrez Gene ID
100755009
Gene Name
protocadherin gamma subfamily A, 9
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614499: 567256 - 572485 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508199.1 XP_003508247.1 1 - 2 2410 99

View Full Product Details