Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fndc8
Entrez Gene ID
100766468
Gene Name
fibronectin type III domain containing 8
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613601: 4533853 - 4538074 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003495802.1 XP_003495850.1 2 - 3 586 75

View Full Product Details