Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
P2ry4
Entrez Gene ID
100769419
Gene Name
pyrimidinergic receptor P2Y, G-protein coupled, 4
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614651: 171894 - 172979 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509114.1 XP_003509162.1 1 - 1 822 68

View Full Product Details