Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kcnc4
Entrez Gene ID
100761530
Gene Name
potassium voltage-gated channel, Shaw-related subfamily, member 4
Gene Aliases
I79_023834
Chromosome Location
Chr.Un|NW_003616747: 22668 - 42417 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007654667.1 XP_007652857.1 3 - 4 1317 62

View Full Product Details