Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccl17
Entrez Gene ID
100764062
Gene Name
chemokine (C-C motif) ligand 17
Gene Aliases
I79_010487
Chromosome Location
Chr.Un|NW_003613984: 1426077 - 1442971 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503627.2 XP_003503675.1 2 - 3 1729 73
XM_007644213.1 XP_007642403.1 3 - 4 469 73
XM_007644214.1 XP_007642404.1 3 - 4 328 73
XM_007644215.1 XP_007642405.1 2 - 3 233 73

View Full Product Details