Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrfn5
Entrez Gene ID
100765666
Gene Name
leucine rich repeat and fibronectin type III domain containing 5
Gene Aliases
I79_022063
Chromosome Location
Chr.Un|NW_003615750: 67511 - 78996 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007653472.1 XP_007651662.1 1 - 2 1377 82

View Full Product Details