Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hsf5
Entrez Gene ID
100758083
Gene Name
heat shock transcription factor family member 5
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614004: 285706 - 332908 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503812.2 XP_003503860.1 3 - 4 214 118

View Full Product Details