Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Frmd1
Entrez Gene ID
100750569
Gene Name
FERM domain containing 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614028: 750713 - 779047 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003504063.1 XP_003504111.1 7 - 8 805 88

View Full Product Details