Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wnt7a
Entrez Gene ID
100772978
Gene Name
wingless-type MMTV integration site family, member 7A
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613584: 1651581 - 1693501 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007612039.1 XP_007610229.1 2 - 3 497 78

View Full Product Details