Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Treml2
Entrez Gene ID
100754919
Gene Name
triggering receptor expressed on myeloid cells-like 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615227: 45077 - 53326 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003511754.1 XP_003511802.1 4 - 5 859 68

View Full Product Details