Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cfl1
Entrez Gene ID
100760435
Gene Name
cofilin 1 (non-muscle)
Gene Aliases
Cofilin-1
Chromosome Location
Chr.Un|NW_003615656: 3608 - 6965 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512921.2 XP_003512969.1 2 - 3 452 75
GenBank mRNA JI890017.1 - 2 - 3 420 75
JP064314.1 - 2 - 3 420 75

View Full Product Details