Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klf8
Entrez Gene ID
100765939
Gene Name
Kruppel-like factor 8
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614419: 255877 - 296111 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007648323.1 XP_007646513.1 5 - 6 1551 96
XM_007648324.1 XP_007646514.1 5 - 6 1031 96
XM_007648325.1 XP_007646515.1 4 - 5 1372 96

View Full Product Details