Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Usp26
Entrez Gene ID
100763595
Gene Name
ubiquitin specific peptidase 26
Gene Aliases
I79_013383
Chromosome Location
Chr.Un|NW_003614212: 939693 - 942221 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506106.1 XP_003506154.1 1 - 1 2219 78

View Full Product Details