Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Emr1
Entrez Gene ID
100751559
Gene Name
egf-like module containing, mucin-like, hormone receptor-like 1
Gene Aliases
I79_008232
Chromosome Location
Chr.Un|NW_003613861: 1642401 - 1715048 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007642457.1 XP_007640647.1 3 - 4 336 107

View Full Product Details