Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Shisa4
Entrez Gene ID
100763467
Gene Name
shisa family member 4
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613706: 1219644 - 1222421 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007634404.1 XP_007632594.1 3 - 4 536 84

View Full Product Details