Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gfral
Entrez Gene ID
100751689
Gene Name
GDNF family receptor alpha like
Gene Aliases
I79_000117
Chromosome Location
Chr.Un|NW_003613582: 422234 - 467793 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003495051.1 XP_003495099.1 3 - 4 303 123

View Full Product Details