Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ptchd3
Entrez Gene ID
100752652
Gene Name
patched domain containing 3
Gene Aliases
I79_018219
Chromosome Location
Chr.Un|NW_003614801: 3765 - 22011 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509990.2 XP_003510038.1 2 - 3 2667 94

View Full Product Details