Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ctdsp2
Entrez Gene ID
100765556
Gene Name
CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) small phosphatase 2
Gene Aliases
I79_015213
Chromosome Location
Chr.Un|NW_003614393: 740046 - 768037 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007648116.1 XP_007646306.1 8 - 9 969 88

View Full Product Details