Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prg2
Entrez Gene ID
100757904
Gene Name
proteoglycan 2, bone marrow (natural killer cell activator, eosinophil granule major basic protein)
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614216: 694175 - 696361 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506160.2 XP_003506208.1 3 - 4 485 81

View Full Product Details