Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klf11
Entrez Gene ID
100753145
Gene Name
Kruppel-like factor 11
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614717: 385363 - 395092 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007650065.1 XP_007648255.1 2 - 3 2452 65
GenBank mRNA JI896543.1 - 2 - 3 1183 65
JP040836.1 - 2 - 3 1183 65

View Full Product Details