Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rgag1
Entrez Gene ID
100759351
Gene Name
retrotransposon gag domain containing 1
Gene Aliases
I79_016757
Chromosome Location
Chr.Un|NW_003614579: 18704 - 59695 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508757.2 XP_003508805.2 3 - 4 4731 95
XM_007649378.1 XP_007647568.1 3 - 4 4693 95

View Full Product Details