Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fut11
Entrez Gene ID
100758336
Gene Name
fucosyltransferase 11 (alpha (1,3) fucosyltransferase)
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613632: 113424 - 117807 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496723.2 XP_003496771.2 1 - 2 815 56

View Full Product Details