Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Comtd1
Entrez Gene ID
100762102
Gene Name
catechol-O-methyltransferase domain containing 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613632: 1264073 - 1266108 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496735.2 XP_003496783.1 3 - 4 749 62

View Full Product Details